Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160167.1 | NP_001153639.1 | 8 | 1006 | ||
NM_001160168.1 | NP_001153640.1 | 5 | 405 | |||
NM_024841.4 | NP_079117.3 | 9 | 992 | |||
GenBank mRNA | AK024275.1 | - | 5 | 352 | ||
AK055042.1 | - | 8 | 893 | |||
BX640959.1 | - | 9 | 973 | |||
DA449593.1 | - | 1 | 41 |