Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024953.3 | NP_079229.2 | 3 | 219 | ||
XM_005253938.1 | XP_005253995.1 | 3 | 184 | |||
XM_006719606.2 | XP_006719669.1 | 3 | 879 | |||
GenBank mRNA | AB054990.1 | - | 3 | 161 | ||
AK292485.1 | - | 3 | 192 | |||
AK307494.1 | - | 4 | 368 | |||
AK309772.1 | - | 3 | 164 | |||
AV760516.2 | - | 3 | 184 | |||
BC034357.1 | - | 3 | 173 | |||
BC065481.1 | - | 3 | 152 | |||
BC113585.1 | - | 3 | 219 | 8322637 | ||
BC113587.1 | - | 3 | 219 | 8322639 | ||
BX641140.1 | - | 3 | 174 | |||
BX647450.1 | - | 3 | 172 |