Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MZT2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025029.3 | NP_079305.2 | 3 | 849 | ||
XM_005263795.4 | XP_005263852.1 | 2 | 754 | |||
XM_011511886.2 | XP_011510188.1 | 4 | 773 | |||
XM_011511887.2 | XP_011510189.1 | 4 | 740 | |||
XM_011511888.2 | XP_011510190.1 | 4 | 1074 | |||
GenBank mRNA | BC014255.2 | - | 3 | 491 | ||
BC066296.1 | - | 3 | 581 | 5284352 |
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FJ224347.1 | - | 517 |