Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025045.5 | NP_079321.3 | 6 | 511 | ||
XM_005261751.4 | XP_005261808.1 | 7 | 512 | |||
XM_011530379.2 | XP_011528681.1 | 6 | 628 | |||
XM_011530380.2 | XP_011528682.1 | 5 | 414 | |||
XM_011530381.2 | XP_011528683.1 | 6 | 405 | |||
XM_011530382.2 | XP_011528684.1 | 6 | 793 | |||
XM_011530383.2 | XP_011528685.1 | 5 | 731 | |||
XM_011530384.2 | XP_011528686.1 | 6 | 669 | |||
GenBank mRNA | AK026235.1 | - | 6 | 469 | ||
AY358164.1 | - | 6 | 511 |