Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025045.5 | NP_079321.3 | 14 | 2084 | ||
XM_005261751.4 | XP_005261808.1 | 15 | 2085 | |||
XM_011530379.2 | XP_011528681.1 | 15 | 2159 | |||
XM_011530380.2 | XP_011528682.1 | 13 | 1987 | |||
XM_011530381.2 | XP_011528683.1 | 14 | 1978 | |||
XM_011530382.2 | XP_011528684.1 | 14 | 2366 | |||
XM_011530383.2 | XP_011528685.1 | 13 | 2304 | |||
XM_011530384.2 | XP_011528686.1 | 14 | 2242 | |||
XM_011530386.2 | XP_011528688.1 | 8 | 1703 | |||
XM_011530387.2 | XP_011528689.1 | 9 | 1488 | |||
XM_011530388.2 | XP_011528690.1 | 8 | 1468 | |||
GenBank mRNA | AK026263.1 | - | 4 | 2008 | ||
AY358164.1 | - | 14 | 2085 |