Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAS1R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138697.3 | NP_619642.2 | 6 | 2446 | ||
NM_177540.2 | NP_803884.1 | 5 | 1684 | |||
XM_011542203.2 | XP_011540505.1 | 6 | 2812 | |||
XM_011542206.2 | XP_011540508.1 | 5 | 2355 | |||
XM_017002402.1 | XP_016857891.1 | 5 | 2998 | |||
XM_017002403.1 | XP_016857892.1 | 4 | 2236 | |||
GenBank mRNA | AF387617.1 | - | 3 | 1791 | ||
AF387618.1 | - | 5 | 2159 | |||
AF387619.1 | - | 4 | 1365 | |||
AK292014.1 | - | 5 | 1653 | |||
AK293499.1 | - | 6 | 1522 | |||
AK315805.1 | - | 6 | 2415 | |||
BC136515.1 | - | 6 | 2446 | 9020505 | ||
BC136516.1 | - | 6 | 2445 | 9020506 |
Set Membership: |
G-protein coupled receptor |