Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284282.1 | NP_001271211.1 | 5 | 1230 | ||
NM_001284283.1 | NP_001271212.1 | 5 | 1164 | |||
NM_001284284.1 | NP_001271213.1 | 5 | 1325 | |||
NM_001321242.1 | NP_001308171.1 | 5 | 1245 | |||
NM_001321243.1 | NP_001308172.1 | 6 | 1398 | |||
NM_001321244.1 | NP_001308173.1 | 6 | 1431 | |||
NM_001321246.1 | NP_001308175.1 | 4 | 1281 | |||
NM_001321247.1 | NP_001308176.1 | 4 | 1165 | |||
NM_001321248.1 | NP_001308177.1 | 4 | 1108 | |||
NM_001321249.1 | NP_001308178.1 | 5 | 1247 | |||
NM_001321250.1 | NP_001308179.1 | 5 | 1269 | |||
NM_001321251.1 | NP_001308180.1 | 5 | 1255 | |||
NM_030796.4 | NP_110423.3 | 5 | 1359 | |||
GenBank mRNA | AB097004.1 | - | 5 | 1218 | ||
AF395824.1 | - | 4 | 1124 | |||
AK075391.1 | - | 6 | 1293 | |||
AK090825.1 | - | 5 | 1269 | |||
AK092992.1 | - | 5 | 1303 | |||
AK126848.1 | - | 1 | 1660 | |||
AK315882.1 | - | 5 | 1157 | |||
AL136610.1 | - | 1 | 1172 | |||
BC016650.1 | - | 1 | 1251 | 4342536 |