Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030931.3 | NP_112193.1 | 2 | 312 | ||
GenBank mRNA | AF525928.1 | - | 2 | 236 | ||
AJ236910.1 | - | 2 | 254 | |||
AK225987.1 | - | 2 | 275 | |||
AY122478.1 | - | 2 | 230 | |||
BX108307.1 | - | 2 | 495 |