Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256139.1 | NP_001243068.1 | 8 | 943 | ||
NM_001256140.1 | NP_001243069.1 | 8 | 1188 | |||
NM_001320732.1 | NP_001307661.1 | 8 | 919 | |||
NM_001320733.1 | NP_001307662.1 | 8 | 880 | |||
NM_001320734.1 | NP_001307663.1 | 7 | 1140 | |||
NM_001747.3 | NP_001738.2 | 8 | 1233 | |||
XM_011533122.1 | XP_011531424.1 | 8 | 1053 | |||
XM_011533123.1 | XP_011531425.1 | 8 | 949 | |||
GenBank mRNA | AK130130.1 | - | 8 | 1233 | ||
AK225374.1 | - | 8 | 861 | |||
AK225378.1 | - | 8 | 861 | |||
AK300507.1 | - | 8 | 825 | |||
AK309290.1 | - | 7 | 602 | |||
AK313898.1 | - | 8 | 887 | |||
BC000728.2 | - | 8 | 887 | 3505088 | ||
BC014549.1 | - | 8 | 889 | 4108934 | ||
BU172582.1 | - | 7 | 792 | |||
FJ224312.1 | - | 8 | 1089 | |||
M94345.1 | - | 8 | 888 |