Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST1H4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003545.3 | NP_003536.1 | 1 | 197 | ||
GenBank mRNA | AK312153.1 | - | 1 | 290 | ||
BC012587.1 | - | 1 | 236 | |||
BC050615.1 | - | 1 | 213 | |||
CR542180.1 | - | 1 | 197 | |||
CR542189.1 | - | 1 | 197 |