Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRISPLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031476.3 | NP_113664.1 | 14 | 1611 | ||
XM_005256190.1 | XP_005256247.1 | 15 | 1866 | |||
GenBank mRNA | AL136861.1 | - | 14 | 1588 | ||
AY358790.1 | - | 14 | 1620 |