Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032289.2 | NP_115665.1 | 10 | 1779 | ||
XM_011537696.2 | XP_011535998.1 | 11 | 1835 | |||
XM_017009975.1 | XP_016865464.1 | 11 | 3142 | |||
XM_017009976.1 | XP_016865465.1 | 11 | 3950 | |||
XM_017009977.1 | XP_016865466.1 | 11 | 3830 | |||
GenBank mRNA | AL136559.1 | - | 10 | 1779 | ||
AL834268.1 | - | 6 | 561 | |||
BC038233.1 | - | 10 | 1756 | 5752229 |