Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALKBH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032306.3 | NP_115682.1 | 4 | 906 | ||
XM_005259658.4 | XP_005259715.1 | 4 | 482 | |||
XM_017027355.1 | XP_016882844.1 | 3 | 763 | |||
GenBank mRNA | AK223336.1 | - | 4 | 529 | ||
AK311956.1 | - | 4 | 545 | |||
AY358858.1 | - | 4 | 906 | |||
AY427650.2 | - | 4 | 906 | |||
BC004393.1 | - | 4 | 561 | 3635540 |