Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288710.1 | NP_001275639.1 | 3 | 450 | ||
NM_032366.4 | NP_115742.3 | 4 | 510 | |||
NR_109976.1 | 3 | 454 | ||||
NR_109977.1 | 3 | 347 | ||||
NR_109978.1 | 4 | 617 | ||||
NR_109979.1 | 2 | 287 | ||||
XM_011522713.2 | XP_011521015.1 | 4 | 709 | |||
GenBank mRNA | AK057077.1 | - | 1 | 1408 | ||
AK293046.1 | - | 1 | 1394 | |||
AK294482.1 | - | 4 | 446 | |||
AW027232.1 | - | 3 | 366 | |||
BG612714.1 | - | 3 | 311 | |||
BI762987.1 | - | 4 | 486 | |||
BP273946.1 | - | 3 | 386 | |||
BQ921757.1 | - | 415 | ||||
BU539308.1 | - | 2 | 235 |