Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C15orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032413.3 | NP_115789.1 | 2 | 319 | ||
NM_197955.2 | NP_922946.1 | 3 | 205 | |||
GenBank mRNA | AB026707.1 | - | 3 | 99 | ||
AF228422.1 | - | 2 | 240 | |||
AK026298.1 | - | 2 | 237 | |||
BC021173.2 | - | 3 | 125 | 5269504 | ||
BM312626.1 | - | 3 | 169 | |||
CA865860.1 | - | 3 | 211 | |||
DB447060.1 | - | 3 | 207 |