Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198777.1 | NP_001185706.1 | 20 | 2238 | ||
NM_001198778.1 | NP_001185707.1 | 20 | 2164 | |||
NM_001198779.1 | NP_001185708.1 | 20 | 2277 | |||
NM_001324375.1 | NP_001311304.1 | 18 | 2041 | |||
NM_001324376.1 | NP_001311305.1 | 21 | 2398 | |||
NM_003591.3 | NP_003582.2 | 20 | 2230 | |||
XM_011519743.1 | XP_011518045.1 | 22 | 2392 | |||
XM_011519744.1 | XP_011518046.1 | 21 | 2274 | |||
XM_011519745.1 | XP_011518047.1 | 21 | 2326 | |||
XM_011519747.1 | XP_011518049.1 | 21 | 2311 | |||
GenBank mRNA | AF126404.1 | - | 19 | 2003 | ||
AK095217.1 | - | 20 | 2238 | |||
AK300491.1 | - | 20 | 2122 | |||
AK310225.1 | - | 21 | 2309 | |||
AL832733.1 | - | 20 | 2214 | |||
BC009591.1 | - | 20 | 2213 | 4104375 | ||
BC110901.1 | - | 20 | 2218 | 6161736 | ||
U58088.1 | - | 17 | 1723 | |||
U83410.1 | - | 20 | 2149 |