Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079692.2 | NP_001073160.1 | 3, 4 | 974 | ||
NM_032597.4 | NP_115986.3 | 4, 5 | 1025 | |||
GenBank mRNA | AF367473.2 | - | 4, 5 | 536 | ||
AK314674.1 | - | 4, 5 | 735 | |||
AY094610.1 | - | 4, 5 | 609 | |||
AY094611.1 | - | 3, 4 | 558 | |||
AY584610.1 | - | 3, 4 | 558 |