Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181704.1 | NP_859055.1 | 396 | |||
GenBank mRNA | AF339515.1 | - | 396 |
Gene Symbol: BAGE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF527552.1 | - | 408 |
Gene Symbol: BAGE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF527550.1 | - | 408 |
Gene Symbol: BAGE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182484.1 | NP_872290.1 | 396 | |||
GenBank mRNA | AF339516.1 | - | 396 |