Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265612.1 | NP_001252541.1 | 22, 23 | 3293 | ||
NM_016284.4 | NP_057368.3 | 22, 23 | 3308 | |||
NM_206999.2 | NP_996882.1 | 22, 23 | 3308 | |||
NR_049763.1 | 22, 23 | 3293 | ||||
GenBank mRNA | AB023224.2 | - | 9, 10 | 1353 | ||
AK056927.1 | - | 13, 14 | 1843 | |||
AL832769.1 | - | 22, 23 | 3257 | |||
AL833549.1 | - | 22, 23 | 3268 | |||
BC024317.1 | - | 5, 6 | 707 | |||
BC040523.1 | - | 22, 23 | 3260 | 5266600 | ||
BX537840.1 | - | 22, 23 | 3239 | |||
BX647815.1 | - | 22, 23 | 3248 | |||
CR749237.1 | - | 15, 16 | 2248 | |||
EF553522.1 | - | 22, 23 | 3242 |