Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142401.1 | NP_001135873.1 | 5 | 610 | ||
NM_001142402.1 | NP_001135874.1 | 5 | 592 | |||
NM_001142403.1 | NP_001135875.1 | 6 | 649 | |||
NM_006016.4 | NP_006007.2 | 6 | 649 | |||
GenBank mRNA | AF106518.1 | - | 5 | 411 | ||
AF263279.1 | - | 6 | 468 | |||
AF299341.1 | - | 6 | 468 | |||
AF299342.1 | - | 5 | 489 | |||
AF299343.1 | - | 5 | 507 | |||
AK301692.1 | - | 5 | 502 | |||
AK303525.1 | - | 5 | 444 | |||
AK308664.1 | - | 5 | 623 | |||
AK310854.1 | - | 6 | 490 | |||
AK312357.1 | - | 6 | 557 | |||
AK315908.1 | - | 5 | 461 | |||
BC011522.2 | - | 6 | 549 | 3854912 | ||
BC037914.1 | - | 5 | 1151 | |||
BC040317.1 | - | 2 | 2501 | |||
D14043.1 | - | 6 | 547 | |||
FJ200494.1 | - | 6 | 464 |