Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C9orf69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256526.1 | NP_001243455.1 | 2 | 1239 | ||
NM_152833.2 | NP_690046.3 | 2 | 1280 | |||
NR_134506.1 | 2 | 1278 | ||||
GenBank mRNA | AK023162.1 | - | 2 | 1231 | ||
AK091349.1 | - | 2 | 1011 | |||
BC013282.2 | - | 2 | 1219 | |||
BC014304.2 | - | 2 | 1177 | |||
BC021231.2 | - | 2 | 1258 | |||
BC064970.1 | - | 1 | 793 | |||
BC084549.1 | - | 1 | 860 | |||
BC092490.1 | - | 2 | 929 |