Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017919.1 | NP_001017919.1 | 8 | 2029 | ||
NM_033544.2 | NP_291022.2 | 9 | 2113 | |||
GenBank mRNA | AK092661.1 | - | 1 | 1544 | ||
AL527085.3 | - | 1 | 388 | |||
BC140709.1 | - | 8 | 2027 | 9021577 |