Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPPLY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138382.2 | NP_612391.1 | 2 | 231 | ||
GenBank mRNA | BC007652.1 | - | 2 | 217 | ||
BC101998.1 | - | 2 | 231 | |||
BC105691.1 | - | 2 | 231 | 40023299 | ||
BC105692.1 | - | 2 | 231 | 40023298 | ||
BC110436.1 | - | 1 | 1148 | 40023301 |