Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATP6AP1L SIRNA Assays ›
Gene Symbol:
ATP6AP1L
Gene Name:
ATPase H+ transporting accessory protein 1 like
Gene Aliases:
-
Chromosome Location:
Chr.5: 82278645 - 82344294 on Build GRCh38
UniGene ID:
Hs.729805
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s228473
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATP6AP1L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001017971.2
NP_001017971.1
9
1681
XM_011543719.2
XP_011542021.1
8
1768
XM_011543720.2
XP_011542022.1
7
1691
XM_011543723.2
XP_011542025.1
9
2328
XM_017010025.1
XP_016865514.1
8
1414
XM_017010026.1
XP_016865515.1
9
1871
XM_017010027.1
XP_016865516.1
9
1219
XM_017010028.1
XP_016865517.1
9
1504
XM_017010029.1
XP_016865518.1
8
1820
XM_017010030.1
XP_016865519.1
8
1453
XM_017010031.1
XP_016865520.1
9
1859
XM_017010032.1
XP_016865521.1
10
2029
XM_017010033.1
XP_016865522.1
9
1492
XM_017010034.1
XP_016865523.1
8
1817
XM_017010035.1
XP_016865524.1
7
1377
XM_017010036.1
XP_016865525.1
8
1792
XM_017010037.1
XP_016865526.1
7
1729
XM_017010038.1
XP_016865527.1
8
1773
XM_017010039.1
XP_016865528.1
4
487
XM_017010040.1
XP_016865529.1
4
392
XM_017010041.1
XP_016865530.1
10
2406
XM_017010042.1
XP_016865531.1
11
2312
XM_017010043.1
XP_016865532.1
10
2395
XM_017010044.1
XP_016865533.1
9
2354
XM_017010045.1
XP_016865534.1
9
1906
XM_017010046.1
XP_016865535.1
9
1987
XM_017010047.1
XP_016865536.1
8
2276
GenBank mRNA
AK022625.1
-
9
1681
AK094973.1
-
2
1574
BC093982.1
-
3
411
7939827
BC112191.1
-
3
411
8327659
Back To Top
More Information
Panther Classification:
Molecular Function -
ATP synthase
Gene Ontology Categories:
Function(s)
Process(es)
ATP hydrolysis coupled proton transport
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-4vtwv:80/10.239.167.181:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline