Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144577.3 | NP_653178.3 | 14 | 3145 | ||
XM_005259413.3 | XP_005259470.1 | 16 | 2853 | |||
XM_005259414.3 | XP_005259471.1 | 16 | 2849 | |||
XM_005259415.3 | XP_005259472.1 | 16 | 2867 | |||
XM_005259416.4 | XP_005259473.1 | 9 | 2000 | |||
XM_011527515.2 | XP_011525817.1 | 17 | 3796 | |||
XM_011527516.2 | XP_011525818.1 | 15 | 2586 | |||
XM_017027483.1 | XP_016882972.1 | 15 | 3626 | |||
GenBank mRNA | AK057357.1 | - | 12 | 1853 | ||
AK057488.1 | - | 8 | 1843 | |||
AK128144.1 | - | 15 | 3209 | |||
AL122083.1 | - | 7 | 1835 | |||
BC007730.2 | - | 14 | 3145 | |||
BC025752.1 | - | 13 | 2662 | |||
BX101994.1 | - | 2 | 320 |