Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORMDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320801.1 | NP_001307730.1 | 6 | 2678 | ||
NM_001320802.1 | NP_001307731.1 | 4 | 1516 | |||
NM_001320803.1 | NP_001307732.1 | 4 | 1230 | |||
NM_139280.3 | NP_644809.1 | 4 | 1521 | |||
XM_017025345.1 | XP_016880834.1 | 4 | 1374 | |||
XM_017025346.1 | XP_016880835.1 | 4 | 1287 | |||
XM_017025347.1 | XP_016880836.1 | 4 | 1467 | |||
XM_017025348.1 | XP_016880837.1 | 4 | 1213 | |||
GenBank mRNA | AF373101.1 | - | 4 | 1257 | ||
AK074811.1 | - | 4 | 1318 | |||
AK075212.1 | - | 4 | 1273 | |||
AX775843.1 | - | 4 | 1275 | |||
BC017087.1 | - | 4 | 1287 | 3850211 | ||
BC071833.1 | - | 4 | 1266 | 6340437 |