Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033258.1 | NP_150283.1 | 2 | 127 | ||
XM_011527524.2 | XP_011525826.1 | 3 | 687 | |||
XM_017027503.1 | XP_016882992.1 | 3 | 266 | |||
XM_017027504.1 | XP_016882993.1 | 3 | 2921 | |||
XM_017027505.1 | XP_016882994.1 | 3 | 220 | |||
GenBank mRNA | AF493875.1 | - | 2 | 127 | ||
BC095514.1 | - | 2 | 130 | 6519176 |