Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171581.1 | NP_001165052.1 | 2, 3 | 656 | ||
NM_004867.4 | NP_004858.1 | 3, 4 | 788 | |||
GenBank mRNA | AB209310.1 | - | 1 | 4616 | ||
AF038953.1 | - | 3, 4 | 590 | |||
AK223327.1 | - | 3, 4 | 567 | |||
AK303238.1 | - | 2, 3 | 438 | |||
AK313154.1 | - | 3, 4 | 746 | |||
AY359051.1 | - | 3, 4 | 529 | |||
BC040437.1 | - | 3, 4 | 543 | 5748519 | ||
BM561934.1 | - | 1 | 31 | |||
CR456875.1 | - | 3, 4 | 451 |