Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAGE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318877.1 | NP_001305806.1 | 4 | 326 | ||
NM_007003.4 | NP_008934.1 | 4 | 338 | |||
GenBank mRNA | AF275258.1 | - | 4 | 324 | ||
AJ005894.1 | - | 4 | 322 | |||
AK312039.1 | - | 4 | 335 | |||
AL548759.3 | - | 3 | 289 | |||
BC010897.2 | - | 3 | 288 | 4043535 | ||
BG435968.1 | - | 4 | 339 | |||
CR456823.1 | - | 3 | 259 | |||
CR542183.1 | - | 3 | 259 | |||
DA841276.1 | - | 4 | 320 |