Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014698.2 | NP_055513.2 | 4 | 821 | ||
XM_006711841.3 | XP_006711904.1 | 4 | 475 | |||
XM_011544328.2 | XP_011542630.1 | 4 | 673 | |||
XM_011544329.2 | XP_011542631.1 | 4 | 1940 | |||
XM_011544330.2 | XP_011542632.1 | 3 | 630 | |||
XM_011544331.2 | XP_011542633.1 | 4 | 504 | |||
XM_011544332.2 | XP_011542634.1 | 4 | 504 | |||
XM_017002916.1 | XP_016858405.1 | 4 | 475 | |||
GenBank mRNA | AB018335.1 | - | 4 | 449 | ||
AK022502.1 | - | 4 | 470 | |||
AK222934.1 | - | 4 | 451 | |||
AK222944.1 | - | 4 | 451 | |||
AK309210.1 | - | 2 | 398 | |||
BC030245.1 | - | 4 | 454 | 5216414 |