Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 13, 14 | 2455 | ||
XM_006721612.2 | XP_006721675.1 | 11, 12 | 1898 | |||
XM_011524090.2 | XP_011522392.1 | 13, 14 | 2455 | |||
XM_011524091.2 | XP_011522393.1 | 13, 14 | 3840 | |||
XM_011524095.2 | XP_011522397.1 | 13, 14 | 2187 | |||
XM_011524096.1 | XP_011522398.1 | 13, 14 | 2455 | |||
XM_011524098.2 | XP_011522400.1 | 11, 12 | 2086 | |||
XM_011524099.2 | XP_011522401.1 | 11, 12 | 1576 | |||
XM_011524100.2 | XP_011522402.1 | 10, 11 | 1358 | |||
XM_017025455.1 | XP_016880944.1 | 13, 14 | 2398 | |||
XM_017025456.1 | XP_016880945.1 | 11, 12 | 2216 | |||
XM_017025457.1 | XP_016880946.1 | 10, 11 | 1986 | |||
GenBank mRNA | AB018296.1 | - | 13, 14 | 2200 | ||
AK292876.1 | - | 13, 14 | 2217 | |||
AK296971.1 | - | 12, 13 | 2097 | |||
AK300547.1 | - | 6, 7 | 603 | |||
AL080108.1 | - | 7, 8 | 888 | |||
BC017566.1 | - | 13, 14 | 2214 | |||
BC113016.1 | - | 12, 13 | 2170 | 40036441 | ||
BC113017.1 | - | 11, 12 | 2063 | 40036446 |