Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015009.2 | NP_055824.1 | 3, 4 | 1022 | ||
XM_017005942.1 | XP_016861431.1 | 2, 3 | 1017 | |||
GenBank mRNA | AB029018.1 | - | 3, 4 | 1022 | ||
BC150623.1 | - | 3, 4 | 1009 | 9056993 | ||
BC152417.1 | - | 3, 4 | 1045 | 8862525 |