Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 18 | 2935 | ||
XM_006721612.2 | XP_006721675.1 | 16 | 2378 | |||
XM_011524090.2 | XP_011522392.1 | 19 | 3001 | |||
XM_011524091.2 | XP_011522393.1 | 19 | 4386 | |||
XM_011524095.2 | XP_011522397.1 | 19 | 2733 | |||
XM_011524096.1 | XP_011522398.1 | 19 | 3001 | |||
XM_011524098.2 | XP_011522400.1 | 17 | 2632 | |||
XM_011524099.2 | XP_011522401.1 | 17 | 2122 | |||
XM_011524100.2 | XP_011522402.1 | 16 | 1904 | |||
XM_017025455.1 | XP_016880944.1 | 19 | 2944 | |||
XM_017025456.1 | XP_016880945.1 | 16 | 2696 | |||
XM_017025457.1 | XP_016880946.1 | 15 | 2466 | |||
GenBank mRNA | AB018296.1 | - | 18 | 2680 | ||
AK292876.1 | - | 18 | 2697 | |||
AK296971.1 | - | 17 | 2577 | |||
AK300547.1 | - | 11 | 1083 | |||
AL080108.1 | - | 12 | 1368 | |||
BC113016.1 | - | 17 | 2650 | 40036441 | ||
BC113017.1 | - | 16 | 2543 | 40036446 |