Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014811.3 | NP_055626.3 | 4 | 4011 | ||
XM_005263411.2 | XP_005263468.1 | 4 | 3816 | |||
XM_017015355.1 | XP_016870844.1 | 4 | 4293 | |||
XM_017015356.1 | XP_016870845.1 | 4 | 4030 | |||
XM_017015357.1 | XP_016870846.1 | 4 | 3884 | |||
XM_017015358.1 | XP_016870847.1 | 4 | 5252 | |||
XM_017015359.1 | XP_016870848.1 | 4 | 4826 | |||
XM_017015360.1 | XP_016870849.1 | 4 | 4836 | |||
XM_017015361.1 | XP_016870850.1 | 3 | 4634 | |||
XM_017015362.1 | XP_016870851.1 | 3 | 4851 | |||
GenBank mRNA | AB014549.1 | - | 4 | 4011 | ||
AK022736.1 | - | 1 | 1746 | |||
BC017511.1 | - | 1 | 1657 | |||
BC047935.1 | - | 1 | 3581 | 5728378 | ||
BC112332.1 | - | 1 | 27 |