Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014865.3 | NP_055680.3 | 5 | 362 | ||
GenBank mRNA | AK022511.1 | - | 5 | 362 | ||
AK303310.1 | - | 4 | 337 | |||
BC016913.1 | - | 5 | 1094 | |||
BC028182.1 | - | 5 | 374 | 5395775 | ||
CN307514.1 | - | 4 | 340 | |||
D63880.1 | - | 5 | 1115 |