Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303139.1 | NP_001290068.1 | 3 | 646 | ||
NM_001303140.1 | NP_001290069.1 | 3 | 509 | |||
NM_001303141.1 | NP_001290070.1 | 3 | 459 | |||
NM_001303142.1 | NP_001290071.1 | 3 | 639 | |||
NM_015009.2 | NP_055824.1 | 5 | 1295 | |||
XM_017005942.1 | XP_016861431.1 | 4 | 1290 | |||
GenBank mRNA | AB029018.1 | - | 5 | 1295 | ||
AK294761.1 | - | 3 | 450 | |||
AK299275.1 | - | 3 | 459 | |||
AK299560.1 | - | 3 | 493 | |||
AK307818.1 | - | 3 | 592 | |||
AK316244.1 | - | 3 | 459 | |||
AK316275.1 | - | 3 | 478 | |||
BC150623.1 | - | 5 | 1282 | 9056993 | ||
BC152417.1 | - | 5 | 1318 | 8862525 | ||
DC412278.1 | - | 3 | 511 |