Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAST3 SIRNA Assays ›
Gene Symbol:
MAST3
Gene Name:
microtubule associated serine/threonine kinase 3
Gene Aliases:
-
Chromosome Location:
Chr.19: 18097764 - 18151692 on Build GRCh38
UniGene ID:
Hs.466184
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22870
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAST3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015016.1
NP_055831.1
24, 25
3080
XM_005259823.3
XP_005259880.1
24, 25
3444
XM_005259824.2
XP_005259881.1
25, 26
3130
XM_005259825.2
XP_005259882.1
25, 26
3127
XM_005259826.3
XP_005259883.1
23, 24
3240
XM_005259827.2
XP_005259884.1
23, 24
3098
XM_006722698.2
XP_006722761.1
24, 25
3122
XM_006722699.2
XP_006722762.2
24, 25
3431
XM_006722700.3
XP_006722763.1
23, 24
3240
XM_011527823.1
XP_011526125.1
26, 27
3220
XM_017026506.1
XP_016881995.1
24, 25
3428
XM_017026507.1
XP_016881996.1
23, 24
3410
XM_017026508.1
XP_016881997.1
23, 24
3408
XM_017026509.1
XP_016881998.1
24, 25
3431
XM_017026510.1
XP_016881999.1
23, 24
3411
XM_017026511.1
XP_016882000.1
25, 26
3518
XM_017026512.1
XP_016882001.1
24, 25
3109
XM_017026513.1
XP_016882002.1
24, 25
3106
XM_017026514.1
XP_016882003.1
25, 26
3113
XM_017026515.1
XP_016882004.1
24, 25
3119
XM_017026516.1
XP_016882005.1
24, 25
3428
GenBank mRNA
AB011133.1
-
24, 25
3078
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
cytoskeleton organization
peptidyl-serine phosphorylation
intracellular signal transduction
magnesium ion binding
protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0