Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161560.2 | NP_001155032.1 | 21 | 2903 | ||
NM_001161561.2 | NP_001155033.1 | 21 | 2840 | |||
NM_001161562.2 | NP_001155034.1 | 20 | 2816 | |||
NM_001161563.2 | NP_001155035.1 | 21 | 2762 | |||
NM_001161564.2 | NP_001155036.1 | 20 | 2738 | |||
NM_001161565.2 | NP_001155037.1 | 20 | 2675 | |||
NM_001161566.2 | NP_001155038.1 | 19 | 2651 | |||
NM_015028.3 | NP_055843.1 | 22 | 2927 | |||
GenBank mRNA | AB011123.2 | - | 22 | 2848 | ||
AB017115.1 | - | 2 | 241 | |||
AF172264.1 | - | 22 | 2582 | |||
AF172265.1 | - | 21 | 2495 | |||
AF172266.1 | - | 21 | 2417 | |||
AF172267.1 | - | 21 | 2558 | |||
AF172268.1 | - | 20 | 2330 | |||
AF172269.1 | - | 20 | 2471 | |||
AF172270.1 | - | 20 | 2393 | |||
AF172271.1 | - | 19 | 2306 | |||
AK291056.1 | - | 20 | 2675 | |||
BC019081.2 | - | 1 | 46 | |||
BC055427.1 | - | 1 | 615 | |||
BC150256.1 | - | 22 | 2848 | 9007184 |