Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161560.2 | NP_001155032.1 | 27 | 3656 | ||
NM_001161561.2 | NP_001155033.1 | 27 | 3593 | |||
NM_001161562.2 | NP_001155034.1 | 26 | 3569 | |||
NM_001161563.2 | NP_001155035.1 | 27 | 3515 | |||
NM_001161564.2 | NP_001155036.1 | 26 | 3491 | |||
NM_001161565.2 | NP_001155037.1 | 26 | 3428 | |||
NM_001161566.2 | NP_001155038.1 | 25 | 3404 | |||
NM_015028.3 | NP_055843.1 | 28 | 3680 | |||
GenBank mRNA | AB011123.2 | - | 28 | 3601 | ||
AF172264.1 | - | 28 | 3335 | |||
AF172265.1 | - | 27 | 3248 | |||
AF172266.1 | - | 27 | 3170 | |||
AF172267.1 | - | 27 | 3311 | |||
AF172268.1 | - | 26 | 3083 | |||
AF172269.1 | - | 26 | 3224 | |||
AF172270.1 | - | 26 | 3146 | |||
AF172271.1 | - | 25 | 3059 | |||
AK026470.1 | - | 2 | 454 | |||
AK291056.1 | - | 26 | 3428 | |||
BC019081.2 | - | 7 | 799 | |||
BC055427.1 | - | 7 | 1368 | |||
BC150256.1 | - | 28 | 3601 | 9007184 |