Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161560.2 | NP_001155032.1 | 29 | 3908 | ||
NM_001161561.2 | NP_001155033.1 | 29 | 3845 | |||
NM_001161562.2 | NP_001155034.1 | 28 | 3821 | |||
NM_001161563.2 | NP_001155035.1 | 29 | 3767 | |||
NM_001161564.2 | NP_001155036.1 | 28 | 3743 | |||
NM_001161565.2 | NP_001155037.1 | 28 | 3680 | |||
NM_001161566.2 | NP_001155038.1 | 27 | 3656 | |||
NM_015028.3 | NP_055843.1 | 30 | 3932 | |||
GenBank mRNA | AB011123.2 | - | 30 | 3853 | ||
AF172264.1 | - | 30 | 3587 | |||
AF172265.1 | - | 29 | 3500 | |||
AF172266.1 | - | 29 | 3422 | |||
AF172267.1 | - | 29 | 3563 | |||
AF172268.1 | - | 28 | 3335 | |||
AF172269.1 | - | 28 | 3476 | |||
AF172270.1 | - | 28 | 3398 | |||
AF172271.1 | - | 27 | 3311 | |||
AK026470.1 | - | 4 | 706 | |||
AK291056.1 | - | 28 | 3680 | |||
BC019081.2 | - | 9 | 1051 | |||
BC055427.1 | - | 9 | 1620 | |||
BC150256.1 | - | 30 | 3853 | 9007184 |