Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF21B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252100.1 | NP_001239029.1 | 6 | 1114 | ||
NM_001252102.1 | NP_001239031.1 | 6 | 1114 | |||
NM_001252103.1 | NP_001239032.1 | 6 | 1114 | |||
NM_017596.3 | NP_060066.2 | 6 | 1114 | |||
GenBank mRNA | AB007918.2 | - | 6 | 1114 | ||
AK131318.1 | - | 3 | 246 | |||
AM177180.1 | - | 6 | 797 | |||
BC137281.1 | - | 6 | 822 | 9021278 | ||
BC144557.1 | - | 6 | 822 | 9053085 |