Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDS5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015032.3 | NP_055847.1 | 12, 13 | 1549 | ||
XM_005266298.4 | XP_005266355.1 | 12, 13 | 1544 | |||
XM_011534999.2 | XP_011533301.1 | 12, 13 | 1544 | |||
XM_011535000.2 | XP_011533302.1 | 12, 13 | 1544 | |||
XM_011535002.2 | XP_011533304.1 | 5, 6 | 646 | |||
XM_017020448.1 | XP_016875937.1 | 12, 13 | 1691 | |||
XM_017020449.1 | XP_016875938.1 | 12, 13 | 1459 | |||
XM_017020450.1 | XP_016875939.1 | 12, 13 | 1459 | |||
XM_017020451.1 | XP_016875940.1 | 12, 13 | 1459 | |||
XM_017020452.1 | XP_016875941.1 | 12, 13 | 1515 | |||
XM_017020453.1 | XP_016875942.1 | 12, 13 | 1544 | |||
GenBank mRNA | AB023196.2 | - | 12, 13 | 1471 | ||
AK299340.1 | - | 12, 13 | 1425 | |||
AL137201.1 | - | 12, 13 | 1489 | |||
BC039256.1 | - | 12, 13 | 1433 | |||
U50533.1 | - | 2, 3 | 300 | |||
U95825.2 | - | 12, 13 | 1428 |