Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001680.4 | NP_001671.2 | 2 | 119 | ||
NM_021603.3 | NP_067614.1 | 2 | 126 | |||
GenBank mRNA | AA309757.1 | - | 2 | 114 | ||
AF241236.1 | - | 2 | 94 | |||
AK096069.1 | - | 2 | 119 | |||
BC005302.1 | - | 2 | 126 | 3828677 | ||
BC013289.1 | - | 2 | 122 | 3686762 | ||
BT006721.1 | - | 2 | 47 | |||
DQ786191.1 | - | 2 | 74 | |||
U50743.1 | - | 2 | 83 |
Gene Symbol: FXYD6-FXYD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204268.1 | NP_001191197.1 | 7 | 732 | ||
NM_001243598.2 | NP_001230527.1 | 7 | 745 | |||
GenBank mRNA | CN310658.1 | - | 7 | 380 | ||
FY211982.1 | - | 7 | 329 | |||
FY214031.1 | - | 7 | 329 |