Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 5 | 492 | ||
NM_001320813.1 | NP_001307742.1 | 5 | 495 | |||
NM_001320814.1 | NP_001307743.1 | 4 | 561 | |||
XM_005270128.3 | XP_005270185.1 | 5 | 473 | |||
XM_005270132.3 | XP_005270189.1 | 4 | 460 | |||
XM_011540124.1 | XP_011538426.1 | 5 | 588 | |||
XM_011540125.1 | XP_011538427.1 | 5 | 531 | |||
XM_017016608.1 | XP_016872097.1 | 4 | 384 | |||
GenBank mRNA | AK225599.1 | - | 5 | 438 | ||
AK298041.1 | - | 5 | 345 | |||
AK298074.1 | - | 4 | 164 | |||
AK298114.1 | - | 5 | 291 | |||
AK313179.1 | - | 5 | 441 | |||
AK316192.1 | - | 5 | 371 | |||
BC111571.1 | - | 5 | 417 | 40069329 | ||
BC139727.1 | - | 3 | 364 | 40024708 | ||
BC157866.1 | - | 5 | 431 | 9057085 | ||
BC157890.1 | - | 5 | 431 | 8862711 | ||
DC358710.1 | - | 4 | 225 | |||
HQ021421.1 | - | 5 | 441 | |||
HQ021422.2 | - | 5 | 441 | |||
HQ021423.2 | - | 5 | 441 | |||
HQ021424.1 | - | 5 | 441 | |||
HQ021425.2 | - | 5 | 441 | |||
HQ021426.1 | - | 5 | 438 | |||
HQ021427.1 | - | 5 | 438 | |||
HQ021428.2 | - | 5 | 438 | |||
HQ021429.2 | - | 5 | 438 | |||
HQ021430.1 | - | 5 | 438 | |||
HQ021431.2 | - | 5 | 438 | |||
HQ021432.1 | - | 5 | 441 | |||
K03475.1 | - | 5 | 429 |