Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM167A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053279.2 | NP_444509.2 | 3 | 1700 | ||
XM_005272398.4 | XP_005272455.1 | 4 | 2053 | |||
XM_011543837.1 | XP_011542139.1 | 3 | 1781 | |||
XM_011543838.2 | XP_011542140.1 | 3 | 1707 | |||
XM_011543840.2 | XP_011542142.1 | 3 | 1597 | |||
GenBank mRNA | AJ301564.1 | - | 3 | 1609 | ||
AL157475.1 | - | 2 | 915 | |||
AL834122.1 | - | 3 | 1689 | |||
BC048214.1 | - | 2 | 889 | |||
BC104041.1 | - | 2 | 1522 | 40026945 | ||
BC104042.1 | - | 2 | 1522 | 40026950 | ||
BC104043.1 | - | 2 | 1522 | 40026951 |