Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099225.1 | NP_001092695.1 | 3 | 376 | ||
NM_144609.2 | NP_653210.2 | 3 | 376 | |||
GenBank mRNA | AK056357.1 | - | 3 | 341 | ||
AL833590.1 | - | 3 | 338 | |||
BC023298.1 | - | 3 | 337 | |||
BC047776.2 | - | 3 | 342 | 6023964 | ||
BG706097.1 | - | 3 | 386 | |||
BX648868.1 | - | 3 | 358 |