Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099225.1 | NP_001092695.1 | 4 | 1413 | ||
NM_144609.2 | NP_653210.2 | 5 | 1472 | |||
GenBank mRNA | AK056357.1 | - | 5 | 1437 | ||
AK098464.1 | - | 1 | 942 | |||
AL833590.1 | - | 4 | 1374 | |||
BC023298.1 | - | 5 | 1433 | |||
BC047776.2 | - | 5 | 1447 | 6023964 | ||
BX648868.1 | - | 4 | 1395 |