Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCOA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318240.1 | NP_001305169.1 | 12 | 6968 | ||
NM_014071.4 | NP_054790.2 | 11 | 4738 | |||
XM_006723755.3 | XP_006723818.1 | 11 | 4666 | |||
XM_011528721.2 | XP_011527023.1 | 11 | 4683 | |||
XM_011528726.2 | XP_011527028.1 | 11 | 4650 | |||
XM_011528727.2 | XP_011527029.1 | 11 | 4647 | |||
XM_017027739.1 | XP_016883228.1 | 12 | 4740 | |||
XM_017027740.1 | XP_016883229.1 | 13 | 4859 | |||
XM_017027741.1 | XP_016883230.1 | 12 | 4740 | |||
XM_017027742.1 | XP_016883231.1 | 12 | 4725 | |||
XM_017027743.1 | XP_016883232.1 | 12 | 4740 | |||
XM_017027744.1 | XP_016883233.1 | 11 | 4629 | |||
XM_017027745.1 | XP_016883234.1 | 11 | 4596 | |||
XM_017027746.1 | XP_016883235.1 | 12 | 4740 | |||
XM_017027747.1 | XP_016883236.1 | 9 | 4117 | |||
XM_017027748.1 | XP_016883237.1 | 12 | 4740 | |||
XM_017027749.1 | XP_016883238.1 | 12 | 4740 | |||
GenBank mRNA | AF128458.1 | - | 10 | 4599 | ||
AF171667.1 | - | 10 | 4541 | |||
AF177388.1 | - | 10 | 4557 | |||
AF208227.1 | - | 12 | 6965 | |||
AF245115.1 | - | 9 | 4456 | |||
AK027040.1 | - | 1 | 694 | |||
BC136272.1 | - | 10 | 4547 | 9020259 | ||
D80003.2 | - | 10 | 4534 | |||
JF707638.1 | - | 9 | 4397 |