Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 2 | 160 | ||
NR_046462.1 | 2 | 295 | ||||
XM_005248480.2 | XP_005248537.1 | 2 | 295 | |||
XM_011543306.2 | XP_011541608.1 | 2 | 382 | |||
XM_011543307.2 | XP_011541609.1 | 2 | 811 | |||
XM_017009332.1 | XP_016864821.1 | 2 | 1316 | |||
XM_017009333.1 | XP_016864822.1 | 2 | 290 | |||
GenBank mRNA | AB020695.1 | - | 2 | 112 | ||
AK290004.1 | - | 2 | 160 | |||
BX640902.1 | - | 2 | 113 | |||
BX647536.1 | - | 2 | 297 | |||
CR936791.1 | - | 3 | 342 | |||
DA326879.1 | - | 2 | 160 | |||
DB024270.1 | - | 2 | 295 |