Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM169A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015566.2 | NP_056381.1 | 6 | 660 | ||
NR_046462.1 | 5 | 623 | ||||
XM_005248480.2 | XP_005248537.1 | 6 | 795 | |||
XM_011543306.2 | XP_011541608.1 | 6 | 882 | |||
XM_011543307.2 | XP_011541609.1 | 6 | 1311 | |||
XM_017009332.1 | XP_016864821.1 | 4 | 1558 | |||
XM_017009333.1 | XP_016864822.1 | 4 | 532 | |||
GenBank mRNA | AB020695.1 | - | 6 | 612 | ||
AK290004.1 | - | 6 | 660 | |||
BX640902.1 | - | 5 | 441 | |||
BX647536.1 | - | 5 | 625 |